siRNA Details
Gene Symbol: Nol8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108408.1 | NP_001101878.1 | 7 | 1347 | ||
XM_006253686.2 | XP_006253748.1 | 7 | 895 | |||
XM_006253687.2 | XP_006253749.1 | 7 | 895 | |||
XM_006253688.2 | XP_006253750.1 | 7 | 1329 | |||
XM_006253689.2 | XP_006253751.1 | 7 | 1329 | |||
XM_006253690.2 | XP_006253752.1 | 7 | 1325 | |||
XM_006253736.2 | XP_006253798.1 | 7 | 895 | |||
XM_006253737.2 | XP_006253799.1 | 7 | 895 | |||
XM_006253738.2 | XP_006253800.1 | 7 | 1342 | |||
XM_006253739.2 | XP_006253801.1 | 7 | 1342 | |||
XM_006253740.2 | XP_006253802.1 | 7 | 1325 |
