siRNA Details
Gene Symbol: RGD1306058 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134572.1 | NP_001128044.1 | 3, 4 | 416 | ||
XM_006253730.2 | XP_006253792.1 | 3, 4 | 399 | |||
XM_008771547.1 | XP_008769769.1 | 2, 3 | 1310 |
