siRNA Details
Gene Symbol: Adtrp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014144.1 | NP_001014166.1 | 1 | 158 | ||
XM_006253833.2 | XP_006253895.1 | 2 | 284 | |||
GenBank mRNA | AY325171.1 | - | 85 | |||
BC088287.1 | - | 1 | 158 | 7321485 |
