siRNA Details
Gene Symbol: Bphl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037206.1 | NP_001032283.1 | 3 | 235 | ||
GenBank mRNA | BC105908.1 | - | 3 | 235 | 7388526 | |
FQ215116.1 | - | 3 | 245 |

More Information
Other Species Targeted

Gene Ontology Categories:
