siRNA Details
Gene Symbol: Dusp22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108412.1 | NP_001101882.1 | 4, 5 | 318 | ||
XM_006253888.2 | XP_006253950.1 | 4, 5 | 266 | |||
XM_006253889.2 | XP_006253951.1 | 4, 5 | 539 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001037955, NM_134068, BC108362, AK149363, BC057944, and others.
Gene Ontology Categories:
