siRNA Details
Gene Symbol: Elmo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108415.1 | NP_001101885.1 | 18, 19 | 2207 | ||
XM_006254087.2 | XP_006254149.1 | 3, 4 | 10820 | |||
XM_008771735.1 | XP_008769957.1 | 18, 19 | 7108 | |||
GenBank mRNA | HQ438489.1 | - | 18, 19 | 1752 |
