siRNA Details
Gene Symbol: Elmo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108415.1 | NP_001101885.1 | 13 | 1508 | ||
XM_008771735.1 | XP_008769957.1 | 13 | 6409 | |||
GenBank mRNA | HQ438489.1 | - | 13 | 1053 |
