siRNA Details
Gene Symbol: Sugct | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014146.1 | NP_001014168.1 | 7 | 692 | ||
GenBank mRNA | BC079348.1 | - | 7 | 692 | 7107655 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
