siRNA Details
Gene Symbol: Svil | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108416.2 | NP_001101886.2 | 8 | 2105 | ||
XM_006254030.2 | XP_006254092.1 | 10 | 2111 | |||
XM_006254031.2 | XP_006254093.1 | 8 | 3849 | |||
XM_008771737.1 | XP_008769959.1 | 10 | 4201 | |||
XM_008771738.1 | XP_008769960.1 | 10 | 1771 | |||
XM_008771739.1 | XP_008769961.1 | 9 | 2002 | |||
XM_008771740.1 | XP_008769962.1 | 9 | 1782 | |||
XM_008771741.1 | XP_008769963.1 | 10 | 2108 | |||
XM_008771742.1 | XP_008769964.1 | 10 | 2111 | |||
XM_008771743.1 | XP_008769965.1 | 10 | 2110 | |||
XM_008771744.1 | XP_008769966.1 | 10 | 2110 | |||
XM_008771745.1 | XP_008769967.1 | 10 | 2107 | |||
XM_008771746.1 | XP_008769968.1 | 10 | 2110 | |||
XM_008771747.1 | XP_008769969.1 | 8 | 1901 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
