siRNA Details
Gene Symbol: Plxdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108422.2 | NP_001101892.1 | 3, 4 | 1249 | ||
GenBank mRNA | BC168868.1 | - | 3, 4 | 1249 | 9088739 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_026162, AK035165, BC057881, AF378761, AK154921, and others.
Gene Ontology Categories:
