siRNA Details
Gene Symbol: Plxdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108422.2 | NP_001101892.1 | 6 | 1534 | ||
GenBank mRNA | BC168868.1 | - | 6 | 1534 | 9088739 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Plxdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108422.2 | NP_001101892.1 | 6 | 1534 | ||
GenBank mRNA | BC168868.1 | - | 6 | 1534 | 9088739 |