siRNA Details
Gene Symbol: Fundc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135757.1 | NP_001129229.1 | 4 | 414 | ||
XM_006254387.2 | XP_006254449.1 | 5 | 974 | |||
XM_006254388.2 | XP_006254450.1 | 4 | 827 | |||
XM_006254389.2 | XP_006254451.1 | 4 | 823 | |||
XM_008771950.1 | XP_008770172.1 | 5 | 970 |
