siRNA Details
Gene Symbol: Fundc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135757.1 | NP_001129229.1 | 4 | 410 | ||
XM_006254387.2 | XP_006254449.1 | 5 | 970 | |||
XM_006254388.2 | XP_006254450.1 | 4 | 823 | |||
XM_006254389.2 | XP_006254451.1 | 4 | 819 | |||
XM_008771950.1 | XP_008770172.1 | 5 | 966 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
