siRNA Details
Gene Symbol: Rnf125 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108424.1 | NP_001101894.1 | 6 | 737 | ||
XM_008771951.1 | XP_008770173.1 | 6 | 660 | |||
GenBank mRNA | BC162061.1 | - | 6 | 710 | 7115974 | |
FQ209520.1 | - | 6 | 957 |

More Information
Other Species Targeted

Gene Ontology Categories:
