siRNA Details
Gene Symbol: Pfdn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108427.1 | NP_001101897.1 | 2 | 200 | ||
XM_006254598.1 | XP_006254660.1 | 2 | 233 | |||
GenBank mRNA | FQ227006.1 | - | 672 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
