siRNA Details
Gene Symbol: Yipf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014150.3 | NP_001014172.3 | 2 | 161 | ||
XM_006254664.2 | XP_006254726.1 | 11 | 1322 | |||
GenBank mRNA | AY321347.1 | - | 11 | 1328 | ||
BC083754.1 | - | 2 | 161 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
