siRNA Details
Gene Symbol: Dtwd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108431.1 | NP_001101901.1 | 4 | 497 | ||
XM_006251008.1 | XP_006251070.1 | 4 | 768 |

More Information
Other Species Targeted

