siRNA Details
Gene Symbol: Snx24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008364.1 | NP_001008365.1 | 5 | 764 | ||
XM_006254726.2 | XP_006254788.1 | 6 | 811 | |||
XM_006254728.1 | XP_006254790.1 | 5 | 519 | |||
XM_006254729.2 | XP_006254791.1 | 6 | 389 | |||
XM_008772152.1 | XP_008770374.1 | 6 | 811 | |||
XM_008772153.1 | XP_008770375.1 | 6 | 770 | |||
GenBank mRNA | BC085883.1 | - | 5 | 764 | 7192093 |
