siRNA Details
Gene Symbol: Adamts19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108433.1 | NP_001101903.1 | 18 | 3177 | ||
XM_006254755.2 | XP_006254817.1 | 18 | 3194 |

More Information
Other Species Targeted

Gene Ontology Categories:
