siRNA Details
Gene Symbol: Cplx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191835.1 | NP_001178764.1 | 1 | 193 | ||
XM_006254878.2 | XP_006254940.1 | 1 | 391 |

More Information
Other Species Targeted

Gene Ontology Categories:
