siRNA Details
Gene Symbol: Hdhd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014151.2 | NP_001014173.2 | 5 | 549 | ||
XM_006254965.2 | XP_006255027.1 | 3 | 260 | |||
XM_006254966.2 | XP_006255028.1 | 3 | 713 | |||
GenBank mRNA | AY539873.1 | - | 5 | 549 | ||
BC078941.1 | - | 3 | 234 | 7109225 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
