siRNA Details
Gene Symbol: Hdhd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006254965.2 | XP_006255027.1 | 2, 3 | 181 | ||
XM_006254966.2 | XP_006255028.1 | 2, 3 | 634 | |||
GenBank mRNA | BC078941.1 | - | 2, 3 | 155 | 7109225 |

More Information
Other Species Targeted

Gene Ontology Categories:
