siRNA Details
Gene Symbol: Brd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108440.1 | NP_001101910.1 | 4 | 334 | ||
XM_006255267.2 | XP_006255329.1 | 5 | 802 | |||
XM_006255268.2 | XP_006255330.1 | 5 | 802 | |||
XM_008772409.1 | XP_008770631.1 | 2 | 408 |
