siRNA Details
Gene Symbol: Phkb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014152.1 | NP_001014174.1 | 30 | 3145 | ||
XM_006255269.2 | XP_006255331.1 | 30 | 3182 | |||
XM_006255270.2 | XP_006255332.1 | 30 | 3182 | |||
XM_008772410.1 | XP_008770632.1 | 30 | 3894 | |||
GenBank mRNA | BC085909.1 | - | 30 | 3145 | 7096506 | |
FQ217825.1 | - | 8 | 867 | |||
FQ234320.1 | - | 5 | 830 |
