siRNA Details
Gene Symbol: Cd97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012164.1 | NP_001012164.1 | 3 | 201 | ||
XM_006255278.2 | XP_006255340.1 | 3 | 886 | |||
XM_006255279.2 | XP_006255341.1 | 3 | 886 | |||
XM_006255280.1 | XP_006255342.1 | 4 | 468 | |||
XM_006255281.2 | XP_006255343.1 | 3 | 886 | |||
GenBank mRNA | BC083857.1 | - | 3 | 201 | 7125718 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
