siRNA Details
Gene Symbol: Lrrc36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004088.3 | NP_001004088.3 | 9 | 1408 | ||
XM_006255503.1 | XP_006255565.1 | 9 | 1396 | |||
XM_006255504.1 | XP_006255566.1 | 9 | 1396 | |||
XM_006255505.1 | XP_006255567.1 | 9 | 1261 | |||
XM_006255507.1 | XP_006255569.1 | 8 | 949 | |||
XM_006255508.2 | XP_006255570.1 | 7 | 1561 | |||
XM_006255509.2 | XP_006255571.1 | 9 | 1337 | |||
GenBank mRNA | AY248698.1 | - | 8 | 1379 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
