siRNA Details
Gene Symbol: Lrrc36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004088.3 | NP_001004088.3 | 14 | 2082 | ||
XM_006255503.1 | XP_006255565.1 | 15 | 2184 | |||
XM_006255504.1 | XP_006255566.1 | 14 | 2064 | |||
XM_006255505.1 | XP_006255567.1 | 15 | 2049 | |||
XM_006255507.1 | XP_006255569.1 | 14 | 1737 | |||
XM_006255508.2 | XP_006255570.1 | 13 | 2349 | |||
XM_006255509.2 | XP_006255571.1 | 15 | 2125 | |||
GenBank mRNA | AY248698.1 | - | 13 | 2040 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
