siRNA Details
Gene Symbol: Ranbp10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135875.1 | NP_001129347.1 | 13 | 1805 | ||
XM_008772517.1 | XP_008770739.1 | 12 | 1820 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
