siRNA Details
Gene Symbol: Cenpn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008366.1 | NP_001008367.1 | 8, 9 | 847 | ||
XM_006255686.2 | XP_006255748.1 | 8, 9 | 961 | |||
XM_006255687.2 | XP_006255749.1 | 8, 9 | 879 | |||
XM_006255689.2 | XP_006255751.1 | 9, 10 | 1183 | |||
XM_006255690.2 | XP_006255752.1 | 9, 10 | 969 | |||
XM_006255691.2 | XP_006255753.1 | 7, 8 | 903 | |||
XM_006255692.2 | XP_006255754.1 | 7, 8 | 783 | |||
GenBank mRNA | AY325257.1 | - | 969 | |||
AY325261.1 | - | 969 | ||||
BC085839.1 | - | 8, 9 | 847 | 7135799 |
