siRNA Details
Gene Symbol: Cenpn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008366.1 | NP_001008367.1 | 6 | 514 | ||
XM_006255686.2 | XP_006255748.1 | 6 | 628 | |||
XM_006255687.2 | XP_006255749.1 | 6 | 546 | |||
XM_006255689.2 | XP_006255751.1 | 7 | 850 | |||
XM_006255690.2 | XP_006255752.1 | 7 | 636 | |||
XM_006255691.2 | XP_006255753.1 | 5 | 570 | |||
XM_006255692.2 | XP_006255754.1 | 5 | 450 | |||
GenBank mRNA | BC085839.1 | - | 6 | 514 | 7135799 |
