siRNA Details
Gene Symbol: Cdh15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207613.1 | NP_997496.1 | 12 | 1964 | ||
GenBank mRNA | AB176538.1 | - | 12 | 1964 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
