siRNA Details
Gene Symbol: Dbndd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014156.1 | NP_001014178.2 | 4 | 595 | ||
GenBank mRNA | BC088277.1 | - | 4 | 595 | 7381798 |

More Information
Other Species Targeted

