siRNA Details
Gene Symbol: Akap7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001801.3 | NP_001001801.1 | 3 | 319 | ||
GenBank mRNA | AY350741.1 | - | 3 | 319 | ||
BC098632.1 | - | 436 | 7315112 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Akap7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001801.3 | NP_001001801.1 | 3 | 319 | ||
GenBank mRNA | AY350741.1 | - | 3 | 319 | ||
BC098632.1 | - | 436 | 7315112 |