siRNA Details
Gene Symbol: Echdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007734.1 | NP_001007735.1 | 6 | 768 | ||
XM_006227746.2 | XP_006227808.1 | 6 | 958 | |||
XM_006227747.2 | XP_006227809.1 | 6 | 1555 | |||
GenBank mRNA | BC079052.1 | - | 6 | 768 | 7115386 |
