siRNA Details
Gene Symbol: Usp29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108465.1 | NP_001101935.1 | 6 | 2351 | ||
XM_006228184.2 | XP_006228246.1 | 5 | 2850 | |||
XM_006228185.2 | XP_006228247.1 | 6 | 2351 |
