siRNA Details
Gene Symbol: Cdc42ep5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006228217.2 | XP_006228279.1 | 2 | 823 | ||
XM_008758944.1 | XP_008757166.1 | 3 | 1110 | |||
XM_008758945.1 | XP_008757167.1 | 2 | 1003 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
