siRNA Details
Gene Symbol: Sult2a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025131.1 | NP_001020302.1 | 1 | 117 | ||
XM_008758949.1 | XP_008757171.1 | 1 | 141 | |||
GenBank mRNA | D14989.1 | - | 1 | 117 | ||
FQ209413.1 | - | 1 | 130 | |||
FQ218086.1 | - | 1 | 104 | |||
FQ219053.1 | - | 1 | 130 | |||
FQ219201.1 | - | 1 | 130 | |||
FQ219357.1 | - | 1 | 103 | |||
FQ219375.1 | - | 1 | 141 | |||
FQ219766.1 | - | 1 | 129 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
