siRNA Details
Gene Symbol: Pnmal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108473.1 | NP_001101943.1 | 2 | 1340 | ||
XM_006228440.2 | XP_006228502.1 | 3 | 1995 | |||
XM_008758951.1 | XP_008757173.1 | 4 | 1711 |
