siRNA Details
Gene Symbol: Pld3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012167.1 | NP_001012167.1 | 9 | 1035 | ||
XM_006228578.2 | XP_006228640.1 | 11 | 1364 | |||
XM_006228579.2 | XP_006228641.1 | 11 | 1312 | |||
XM_006228580.2 | XP_006228642.1 | 10 | 1470 | |||
XM_006228581.2 | XP_006228643.1 | 11 | 1342 | |||
XM_008759165.1 | XP_008757387.1 | 10 | 1255 | |||
GenBank mRNA | BC089987.1 | - | 9 | 1035 | 7318612 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
