siRNA Details
Gene Symbol: Sirt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008368.1 | NP_001008369.1 | 8 | 964 | ||
XM_006228672.2 | XP_006228734.1 | 8 | 696 | |||
XM_006228673.2 | XP_006228735.1 | 9 | 701 | |||
XM_006228675.1 | XP_006228737.1 | 8 | 696 | |||
XM_006228676.2 | XP_006228738.1 | 9 | 701 | |||
GenBank mRNA | BC086545.1 | - | 8 | 964 | 7134377 | |
FQ212805.1 | - | 8 | 623 | |||
FQ216424.1 | - | 4 | 292 |
