siRNA Details
Gene Symbol: Rbm42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014159.2 | NP_001014181.2 | 2 | 338 | ||
XM_006228781.2 | XP_006228843.1 | 2 | 363 | |||
GenBank mRNA | BC079321.1 | - | 2 | 296 | 7135399 | |
BP504208.1 | - | 2 | 343 | |||
FQ222852.1 | - | 2 | 308 |
