siRNA Details
Gene Symbol: Lgi4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199499.1 | NP_955793.1 | 6 | 804 | ||
XM_008759169.1 | XP_008757391.1 | 5 | 1247 | |||
XM_008759170.1 | XP_008757392.1 | 6 | 518 | |||
GenBank mRNA | BC064659.1 | - | 6 | 804 | 5623799 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
