siRNA Details
Gene Symbol: Scgb1b30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100859.1 | NP_001094329.1 | 2 | 83 | ||
XM_008759171.1 | XP_008757393.1 | 2 | 213 | |||
GenBank mRNA | AY871780.1 | - | 1 | 16 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
