siRNA Details
Gene Symbol: Kdelr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017385.1 | NP_001017385.1 | 4 | 572 | ||
GenBank mRNA | BC092600.1 | - | 4 | 572 | 7121304 |

More Information
Other Species Targeted

Gene Ontology Categories:
