siRNA Details
Gene Symbol: Ube3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191837.2 | NP_001178766.1 | 6, 7 | 2272 | ||
XM_006229310.2 | XP_006229372.1 | 5, 6 | 3117 | |||
XM_006229313.2 | XP_006229375.1 | 4, 5 | 2997 | |||
XM_008759399.1 | XP_008757621.1 | 3, 4 | 3827 | |||
XM_008759400.1 | XP_008757622.1 | 5, 6 | 1758 | |||
XM_008759401.1 | XP_008757623.1 | 5, 6 | 3061 |

More Information
Other Species Targeted

Gene Ontology Categories:
