siRNA Details
Gene Symbol: Ube3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191837.2 | NP_001178766.1 | 9 | 2755 | ||
XM_006229310.2 | XP_006229372.1 | 8 | 3600 | |||
XM_006229313.2 | XP_006229375.1 | 7 | 3480 | |||
XM_008759399.1 | XP_008757621.1 | 6 | 4310 | |||
XM_008759400.1 | XP_008757622.1 | 8 | 2241 | |||
XM_008759401.1 | XP_008757623.1 | 8 | 3544 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_011668, NM_173010, NM_001033962, AK083795, U82122, and others.
Gene Ontology Categories:
