siRNA Details
Gene Symbol: Lrrc28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108486.1 | NP_001101956.1 | 2 | 317 | ||
XM_006229346.2 | XP_006229408.1 | 2 | 371 | |||
XM_006229347.1 | XP_006229409.1 | 2 | 332 | |||
GenBank mRNA | FQ212061.1 | - | 2 | 351 |
