siRNA Details
Gene Symbol: Lrrc28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108486.1 | NP_001101956.1 | 9 | 1072 | ||
XM_006229346.2 | XP_006229408.1 | 9 | 1126 | |||
XM_006229347.1 | XP_006229409.1 | 8 | 880 | |||
GenBank mRNA | FQ212061.1 | - | 9 | 1010 | ||
FQ222679.1 | - | 2 | 214 |
