siRNA Details
Gene Symbol: Me3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108491.2 | NP_001101961.2 | 15 | 1792 | ||
XM_006229666.1 | XP_006229728.1 | 14 | 1690 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
