siRNA Details
Gene Symbol: Fam168a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108494.1 | NP_001101964.1 | 5 | 505 | ||
XM_006229773.2 | XP_006229835.1 | 4 | 1662 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
